missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ABpure™ Antibody Purification Kit
Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
Description
- Used to purify antibodies from crude samples such as ascites fluid or immune serum
- Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
- 20 to 500μg of antibody can be purified in each run
Specifications
Specifications
| Content And Storage | 4°C |
| Quantity | 1 Test |
| Kit Contents | 4 or 12 additional collecting tubes, 1 vial AbPure wash buffer, 1 vial AbPure eution buffer, 1 vial AbPure neutralization buffer, 1 or 3 vials Protein A resin, 1 vial 10X AbPure binding buffer, 1/3 spin cartridge/collecting tube assemblies |
| Sufficient For | 1 Purification |
| Product Type | Antibody Purification Kit |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction