missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ABpure™ Mouse Antibody Purification Kit

Product Code. 18093544 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
18093544 1 Purification 1 test
18061004 3 Purifications 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Not suitable for use with antibodies from other species
Product Code. 18093544 Supplier Novus Biologicals™ Supplier No. 2610005
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Not suitable for use with antibodies from other species

Novus Biologicals™ ABpure Mouse Antibody Purification Kit is used to remove contaminants such as BSA and other proteins, glycine, tris, and azide that interfere in conjugation reactions.

  • Used to purify antibodies from crude samples such as ascites fluid or immune serum
  • Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
  • 20 to 150μg of antibody can be purified in each run
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Quantity 1 Test
Kit Contents 4 or 12 additional collecting tubes, 1 vial AbPure elution buffer, 1 vial AbPure wash buffer, 1 vial AbPure neutralization buffer, 1 vial 10x AbPure binding buffer, 1 or 3 spin cartridge/collecting tube assemblies, 1 or 3 vials of AbPure Mouse resin
Sufficient For 1 Purification
Product Type Antibody Purification Kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.