missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Fully compatible with InnovaCoat GOLD and LATEX conjugation kits which allow purified antibody to be conjugated rapidly
- Antibody to be purified should be in 10mL to 50mL of tissue culture supernatant (TCS)
- Up to 5mg of antibody can be purified in each run
Specifications
Specifications
| Sufficient For | 3 Purifications |
| Product Type | Antibody Purification Kit |
| Content And Storage | 1 or 3 spin purification columns with caps and stoppers, 1 or 3 concentrator spin columns, 1 or 3 bottles AbPure TCS Protein A resin, 1 bottle 10x AbPure binding buffer, 1 bottle AbPure preservative, 1 bottle AbPure wash buffer, 1 bottle AbPure elution buffer. 4°C |
| Product Line | Novus |
| Quantity | 3 Tests |
For research use only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?