missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AbSelect™ TCS Purification System Antibody Purification Kit

Product Code. 18004365 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Test
Unit Size:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18004365 1 Test 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18004365 Supplier R&D Systems™ Supplier No. 8620030
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Fast and easy to use kit for purification of IgG fractions from hybridoma supernatants

AbSelect TCS is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from hybridoma supernatants. The method involves the capture of the antibody on the AbSelect TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.

  • Purify antibody from 10 to 50mL of hybridoma TCS
  • 100% compatible with lightning-link range of conjugation kits
  • Supplied with all components and buffers
  • Available with full technical support

Recommended Storage

Store at 4°C. Do not freeze

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Purification Kit
Product Line Novus
For Use With (Application) The antibody to be purified should be in 10 to 50 mL of tissue culture supernatant. Up to 5 mg of antibody can be purified in each run.
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.