Learn More
Description
The AbSelect™ Rat TCS Antibody Purification System involves the capture of the antibody on the AbSelect™ Rat resin and the removal of unwanted substances using a simple wash procedure. The purified antibody is then eluted and neutralized. AbSelect™ Rat TCS Antibody Purification System is not suitable for use with antibodies from other species.
The components of all AbSelect™ kits are fully compatible with the Lightning-Link™ and Lightning-Link™ antibody labeling systems (available separately), which allow the purified antibody to be rapidly conjugated to a wide variety of labels.
Kit Contents:
- 1 vial of Elution Buffer
- 1 vial of 10x Binding Buffer
- 12 additional collecting tubes
- 1 vial of Neutralization Buffer
- 1 vial of Wash Buffer
- 3 concentrator spin columns
- 3 vials of AbSelectTM Rat resin
- 3 spin cartridge/ collecting tube assemblies
Specifications
Specifications
| Content And Storage | 4°C |
| Quantity | 3 Tests |
| Target | AbSelect (TM) Rat TCS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.