Learn More
ACAA1 Antibody (CL2660) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42383FR
Product Code. 30287431
  • 737.00 € / 0.10mL
Description
ACAA1 Monoclonal antibody specifically detects ACAA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACAA1
Monoclonal
DyLight 680
Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 1, EC 2.3.1, peroxisomal
This antibody was developed using a recombinant protein derived from P09110, with the exact immunogen sequence remaining proprietary.
0.1 mL
Proteases & Other Enzymes, Signal Transduction
30
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL2660
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified