missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAA2 Antibody (OTI1C10), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71602PE
This item is not returnable.
View return policy
Description
ACAA2 Monoclonal antibody specifically detects ACAA2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| ACAA2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI1C10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| 3-ketoacyl-CoA thiolase, mitochondrial, Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 2, acetyl-Coenzyme A acyltransferase 2, beta ketothiolase, beta-ketothiolase, DSAEC, EC 2.3.1, EC 2.3.1.16, FLJ35992, FLJ95265, Mitochondrial 3-oxoacyl-CoA thiolase, mitochondrial 3-oxoacyl-Coenzyme A thiolase, T1 | |
| Full length human recombinant protein of human ACAA2(NP_006102) produced in HEK293T cell. | |
| 0.1 mL | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 10449 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction