missing translation for 'onlineSavingsMsg'
Learn More

Acacetin, MedChemExpress

Product Code. 16468988
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16468988

Brand: Medchem Express HYN0451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Acacetin (5,7-Dihydroxy-4'-methoxyflavone) is an orally active flavonoid derived from Tephroseris kirilowii (Turcz.) Holub. Acacetin docks in the ATP binding pocket of PI3Kγ. Acacetin causes cell cycle arrest and induces apoptosis and autophagy in cancer cells. Acacetin has potent anti-cancer and anti-inflammatory activity and has the potential for pain-related diseases research.

Acacetin (5,7-Dihydroxy-4'-methoxyflavone) is an orally active flavonoid derived from Tephroseris kirilowii (Turcz.) Holub. Acacetin docks in the ATP binding pocket of PI3Kγ. Acacetin causes cell cycle arrest and induces apoptosis and autophagy in cancer cells. Acacetin has potent anti-cancer and anti-inflammatory activity and has the potential for pain-related diseases research.

Specifications

Chemical Name or Material Acacetin
CAS 480-44-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12O5
Quantity 10 mg
Synonym 5,7-Dihydroxy-4'-methoxyflavone
Solubility Information DMSO : ≥ 37 mg/mL (130.16 mM)
SMILES O=C1C=C(C2=CC=C(OC)C=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 284.26
Formula Weight 284.26
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.