missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 549.00 €
Specifications
| Antigen | ACADS |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18494811
|
Novus Biologicals
NBP1-89290-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263188
|
Novus Biologicals
NBP1-89290 |
0.1 mL |
549.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACADS Polyclonal specifically detects ACADS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ACADS | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ACAD3, acyl-CoA dehydrogenase, C-2 to C-3 short chain, acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain, Butyryl-CoA dehydrogenase, EC 1.3.99, EC 1.3.99.2, mitochondrial short-chain specific acyl-CoA dehydrogenase, SCADshort-chain acyl-CoA dehydrogenase, short-chain specific acyl-CoA dehydrogenase, mitochondrial, unsaturated acyl-CoA reductase | |
| ACADS | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 35 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ELPETHQMLLQTCRDFAEKELFPIAAQVDKEHLFPAAQVKKMGGLGLLAMDVPEELGGAGLDYLAYAIAMEEISRGCASTGVIMSVNNSLYLGPILKFGSKEQKQAWVTPFTSGDKIGCFALSEPGNGS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title