missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADVL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89287
This item is not returnable.
View return policy
Description
ACADVL Polyclonal specifically detects ACADVL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ACADVL | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50-1:200 | |
| ACAD6, acyl-CoA dehydrogenase, very long chain, acyl-Coenzyme A dehydrogenase, very long chain, EC 1.3.99, EC 1.3.99.-, LCACD, very long-chain specific acyl-CoA dehydrogenase, mitochondrial, VLCAD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human ACADVL antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ACADVL | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KLIKHKKGIVNEQFLLQRLADGAIDLYAMVVVLSRASRSLSEGHPTAQHEKMLCDTWCIEAAARIREGMAALQSDPWQQELYRNFKSISKALVERGGVVTSNPLGF | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 37 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction