missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
369.00 € - 543.00 €
Specifications
| Antigen | ACAT1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockout Validated, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18412391
|
Novus Biologicals
NBP1-89285-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727003
|
Novus Biologicals
NBP1-89285 |
543.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ACAT1 Polyclonal specifically detects ACAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| ACAT1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL | |
| ACAT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human ACAT1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockout Validated, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 38 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VSATRTPIGSFLGSLSLLPATKLGSIAIQGAIEKAGIPKEEVKEAYMGNVLQGGEGQAPTRQAVLGAGLPISTPCTTINKVCASGMKAIMMASQSLMCGHQDVMVAGGMESMSNVPYVMNRGSTPYGGVKLEDLIVKDGLTD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title