missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT2 Antibody (SR1630), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | ACAT2 |
|---|---|
| Clone | SR1630 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602404
|
Novus Biologicals
NBP3-22462-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660785
|
Novus Biologicals
NBP3-22462-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACAT2 Monoclonal antibody specifically detects ACAT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| ACAT2 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| acetoacetyl Coenzyme A thiolase, acetyl-CoA acetyltransferase 2, acetyl-CoA acetyltransferase, cytosolic, Acetyl-CoA transferase-like protein, acetyl-Coenzyme A acetyltransferase 2, acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase), ACTL, Cytosolic acetoacetyl-CoA thiolase, EC 2.3.1, EC 2.3.1.9 | |
| A synthesized peptide derived from human ACAT2 (Uniprot #: Q9BWD1) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1630 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 39 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title