Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator
Shop All Fisherbrand Products
Click to view available options
Holds:
1.5/2.0 mL Tubes
10-15 mL and 5-7 mL Tubes
14 mm dia. Culture Tubes
15 mL Tubes
18.5 mm dia. Culture Tubes
50 mL Tubes
Unit Size:
Each
Description
Multi-purpose tube rotator offers flexibility with multiple carousel options
- Tube rotator
Specifications
Specifications
| Holds | 10-15 mL and 5-7 mL Tubes |
| Product Type | Tube Carousel |
| No. of Positions | 10, 20 |