Learn More
Fisherbrand™ Accessories for Incubating Mini Shaker
Product Code. 15594100 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Centrifuge Tube Rack
Flask Clamp
Sticky Mat
Tube Rack with Adapter
Holds:
1.5 - 2.0mL Tubes
100, 125 mL Flasks
15 mL Centrifuge Tubes
25 mL Flasks
250 mL Flasks
50 mL Centrifuge Tubes
50 mL Flasks
N/A
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Holds unitSize
15594100 Flask Clamp 250 mL Flasks Each
15554110 Sticky Mat N/A Each
15564100 Flask Clamp 25 mL Flasks Each
15574080 Centrifuge Tube Rack 50 mL Centrifuge Tubes Each
15574100 Flask Clamp 50 mL Flasks Each
15564080 Centrifuge Tube Rack 15 mL Centrifuge Tubes Each
15584100 Flask Clamp 100, 125 mL Flasks Each
15554080 Tube Rack with Adapter 1.5 - 2.0mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15594100
Add to basket
Add to basket
Accessories required for the use of Fisher Scientific™ Incubating Mini Shaker
Accessories for incubating mini shaker
  • Increase performance and flexibility
TRUSTED_SUSTAINABILITY
Specifications
Holds 250 mL Flasks
Product Type Flask Clamp
Material Stainless Steel
No. of Positions 8