missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ PepMap™ 100 C18 HPLC Columns

Product Code. 11352013 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
5 mm
15 mm
20 mm
50 mm
150 mm
250 mm
500 mm
750 mm
Diameter (Metric):
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.3 mm
0.75 mm
75 μm
1 mm
Particle Size:
2 μm
3 μm
5 μm
Product Type:
Analytical Column
Analytical Column, NanoViper
Analytical nano Column
Column
Trap Cartridge
Trap Column
Unit Size:
Each
Pack of 2
Pack of 5
39 product options available for selection
Product selection table with 39 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Product Type Unit Size
11352013 150 mm 0.075 mm 3 μm Trap Column Pack of 2
11342263 50 mm 0.075 mm 3 μm Analytical Column, NanoViper Each
11352263 150 mm 0.075 mm 3 μm Analytical Column Each
11362263 250 mm 0.075 mm 3 μm Analytical Column Each
11372263 500 mm 0.075 mm 3 μm Analytical Column Each
11302273 150 mm 1 mm 3 μm Analytical Column Each
11312113 5 mm 1 mm 5 μm Trap Cartridge Each
11362113 5 mm 0.3 mm 5 μm Trap Cartridge Pack of 5
11322113 15 mm 1 mm 5 μm Trap Cartridge Each
11382243 150 mm 0.1 mm 5 μm Trap Column Pack of 2
11332253 150 mm 0.2 mm 5 μm Trap Column Pack of 2
11392243 150 mm 0.1 mm 5 μm Trap Column Pack of 2
11352103 150 mm 0.075 mm 5 μm Analytical Column Each
11362103 250 mm 0.075 mm 5 μm Analytical Column Each
11352023 150 mm 0.05 mm 2 μm Analytical Column, NanoViper Each
11372013 150 mm 0.3 mm 2 μm Analytical Column Each
11392253 50 mm 0.75 mm 2 μm Analytical Column, NanoViper Each
11382013 500 mm 0.75 mm 2 μm Analytical Column Each
11342013 150 mm 0.075 mm 2 μm Analytical Column Each
11342023 50 mm 0.05 mm 2 μm Analytical Column, NanoViper Each
11332023 50 mm 0.3 mm 2 μm Analytical Column Each
15597520 250 mm 0.75 mm 2 μm Analytical Column Each
15810705 500 mm 0.075 mm 2 μm Analytical Column Each
15820705 500 mm 0.075 mm 3 μm Analytical Column Each
15567530 500 mm 0.075 mm 2 μm Analytical Column Each
15572920 750 mm 0.075 mm 2 μm Analytical nano Column Each
15800705 50 mm 1 mm 2 μm Analytical Column, NanoViper Each
15588090 20 mm 75 μm 3 μm Trap Column Each
15527660 150 mm 0.05 mm 2 μm Analytical Column Each
12739835 150 mm 0.05 mm 2 μm Analytical Column Each
11382253 250 mm 0.075 mm 3 μm Analytical Column Each
11342103 150 mm 0.075 mm 3 μm Analytical Column Each
11312263 150 mm 0.1 mm 5 μm Trap Column Pack of 2
15517460 150 mm 0.075 mm 2 μm Analytical Column Each
17294434 150 mm 0.1 mm 5 μm Trap Column Pack of 2
17204444 150 mm 0.3 mm 3 μm Analytical Column, NanoViper Each
13486836 150 mm 1 mm 2 μm Analytical Column Each
15518080 250 mm 0.05 mm 2 μm Column Each
15588080 500 mm 0.05 mm 2 μm Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
39 options
This item is not returnable. View return policy
Product Code. 11352013 Supplier Thermo Scientific™ Supplier No. 164535
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 30-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for high-resolution tryptic, natural, and synthetic peptide analyses. Its high loading capacity is exceptional for low abundant peptides in complex proteomic samples.

Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ PepMap™ 100 C18 LC Columns. PepMap LC columns offer lengths targeting fast or high resolution peptide separation.

  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows highest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Preassembled with nanoViper column inlet fittings for easy column installation

Key advantages of nanoViper fittings

  • Ease of use, no sleeve for small capillaries
  • Tool-free assembly
  • No column damage from overtightening: capable of withstanding pressures up to 1000 bars and is compatible with third-party valves and unions
  • No experimental failure due to bad connections

Ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides as well as LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.

  • High loading capacity
  • Use for the analysis of low abundant peptides often found in complex proteomics samples
  • Select the column length and I.D. for your throughput, sensitivity, and amount of sample available requirements
  • Choose from 50, 150, 250, 500mm lengths and 50, 75, 300 and 1000μm I.D.
  • Superior protein and peptide separations
  • Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
  • Outstanding separation efficiency
  • Superior resolution and recovery
  • Easy installation with nanoViper™ Fingertight Fittings

PepMap nanocolumns are also available pre-assembled with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fitting for a dead-volume-free connection to 1000 bar.

  • Requires no tools
  • Provides virtually zero-dead-volume connections
  • Withstands pressures up to a 1000 bar

About Nano, capillary, and micro Acclaim PepMap columns

Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and microcolumns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and microcolumn in a variety of chemistries for peptides and protein analysis.

Use PepMap Trap columns for desalting and preconcentration of peptides before LC separation with MS detection. The trap columns provide the highest efficiencies for one-dimensional peptide mapping experiments and 2D-LC analyses.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Trap Column, NanoViper
Diameter (Metric) 0.075 mm
For Use With UltiMate 3000 RSLC Nano System
Length (Metric) 150 mm
Max. Pressure 7250 psi (500 bar)
Particle Size 3 μm
pH 2 to 8
Pore Size 100 Å
Quantity Pack of 2
Surface Area 450 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line nanoLC
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Material PEEK Shielded Fused Silica
Bed Length 20 mm
Product Type Trap Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.