missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ C30 HPLC Columns

Product Code. 11312003 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Particle Size:
3 μm
5 μm
Packungsgröße:
Each
Pack of 2
Artikelnummer. Length (Metric) Diameter (Metric) Particle Size unitSize
11312003 250 mm 4.6 mm 5 μm Each
11322003 150 mm 4.6 mm 5 μm Each
12769815 100 mm 4.6 mm 3 μm Each
12779815 50 mm 4.6 mm 3 μm Each
12799815 50 mm 3 mm 3 μm Each
12709825 250 mm 2.1 mm 3 μm Each
12719825 100 mm 2.1 mm 3 μm Each
12729825 50 mm 2.1 mm 3 μm Each
11332003 10 mm 4.6 mm 5 μm Pack of 2
11342003 10 mm 3 mm 5 μm Pack of 2
11352003 10 mm 2.1 mm 5 μm Pack of 2
11362003 150 mm 4.6 mm 3 μm Each
11372003 150 mm 3 mm 3 μm Each
11362023 150 mm 2.1 mm 3 μm Each
11372023 250 mm 3 mm 3 μm Each
15 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11312003

Marke: Thermo Scientific™ 075718

Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Use for superior separation of hydrophobic structural isomers.

For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim™ C30 LC reversed-phase columns provide superior results. The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.

High Shape Selectivity for the Separation of Structurally Related Isomers
  • Unique selectivity complementary to other reversed-phase columns
  • High efficiency and rugged packing
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8
  • Available in 3 and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 200Å
About the Acclaim Family of HPLC Columns

Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.

  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
TRUSTED_SUSTAINABILITY

Spezifikation

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Max. Pressure 8000 psi (550 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 200 Å
Quantity Each
Surface Area 200 m²/g
Temperature 60°C
USP Type L62
Stationary Phase C30
Product Line Acclaim
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 13%
Column Type Reversed Phase
Endcapped Proprietary
Phase Reversed Phase
Green Features Responsibly packaged
Mehr anzeigen Weniger anzeigen

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.