missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ SEC-300 Size Exclusion Chromatography HPLC Columns

Product Code. 15178744 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
33 mm
150 mm
300 mm
Diameter (Metric):
4.6 mm
7.8 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
15178744 300 mm 7.8 mm Each
15108754 300 mm 4.6 mm Each
15128754 150 mm 7.8 mm Each
15148754 33 mm 4.6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15178744 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 079725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.

Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC-300 Size Exclusion Chromatography HPLC Columns. These resin-based, high-performance, size-exclusion chromatography columns cover molecular weight range: 5μm, 300Å. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.

The Acclaim SEC-300 column is calibrated from 100 to 50,000 Daltons and offers good resolution for lower molecular weight polymers or oligomers.

High-Performance Analysis of Water-Soluble Polymers
  • High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
  • Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 7.8 mm
Flow Rate 1 mL/min
Length (Metric) 300 mm
Max. Pressure 1200 psi (80 bar)
Particle Shape Spherical
Particle Size 5 μm
pH 2 to 12
Pore Size 300 Å
Quantity Each
Temperature 60°C
USP Type N/A
Stationary Phase Proprietary
Product Line Acclaim
Packing Material Spherical, Fully Porous Polymer
Column Type Size Exclusion
Endcapped Yes
Phase Size Exclusion
Technique Bio-LC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.