missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE2 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80063-1-RR-20UL
This item is not returnable.
View return policy
Description
The ACE2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human ACE2. This antibody recognizes human antigen. The ACE2 antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.Specifications
| ACE2 | |
| Recombinant Monoclonal | |
| 500 μg/mL | |
| ACEH; DKFZp434A014 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Recombinant Antibody | |
| IgG |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 4H12 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 59272 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction