missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol hydrochloride, MedChemExpress

Product Code. 16437336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437336 100 mg 100mg
16437368 10 g 10g
16429847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16437336 Supplier Medchem Express Supplier No. HY17497A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.

Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.

Specifications

Chemical Name or Material Acebutolol hydrochloride
CAS 34381-68-5
Color White
Purity Grade Notes Research
Molecular Formula C18H29ClN2O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (268.18 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (134.09 mM)
SMILES CCCC(NC1=CC=C(OCC(O)CNC(C)C)C(C(C)=O)=C1)=O.Cl
Molecular Weight (g/mol) 372.89
Formula Weight 372.89
Percent Purity 98.83%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.