missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol hydrochloride, MedChemExpress

Product Code. p-200096389
Encompass_Preferred
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
This item is not returnable. View return policy

Product Code. 16437368

Brand: Medchem Express HY17497A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.

Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.

Specifications

Chemical Name or Material Acebutolol hydrochloride
CAS 34381-68-5
Color White
Purity Grade Notes Research
Molecular Formula C18H29ClN2O4
Quantity 10 g
Solubility Information DMSO : 100 mg/mL (268.18 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (134.09 mM)
SMILES CCCC(NC1=CC=C(OCC(O)CNC(C)C)C(C(C)=O)=C1)=O.Cl
Molecular Weight (g/mol) 372.89
Formula Weight 372.89
Percent Purity 98.83%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.