missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aceclofenac, MedChemExpress
Description
Aceclofenac is an orally active nonsteroidal anti-inflammatory drug (NSAID), with analgesic and anti-inflammatory properties. Aceclofenac is used for the research of osteoarthritis, ankylosing spondylitis, rheumatoid arthritis.
Specifications
Specifications
| Chemical Name or Material | Aceclofenac |
| Physical Form | Solid |
| CAS | 89796-99-6 |
| Color | Off-White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C16H13Cl2NO4 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 100 mg/mL (282.34 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic and warming) |
| SMILES | O=C(OCC(O)=O)CC1=CC=CC=C1NC2=C(Cl)C=CC=C2Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?