missing translation for 'onlineSavingsMsg'
Learn More

Acenocoumarol, TRC

Product Code. 18082150
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18082150 25 mg 25mg
18052550 10 mg 10mg
17939969 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18082150 Supplier Toronto Research Chemicals Supplier No. TRCA13080025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-72-7
Molecular Formula C19 H15 N O6
Molecular Weight (g/mol) 353.33
Synonym 2H-1-Benzopyran-2-one, 4-hydroxy-3-[1-(4-nitrophenyl)-3-oxobutyl]-, Acenocoumarol (6CI), Coumarin, 3-(α-acetonyl-p-nitrobenzyl)-4-hydroxy- (8CI), (±)-Acenocoumarin, (±)-Acenocoumarol, (±)-Nicoumalone, (±)-p-Nitrowarfarin, 3-(Alpha-acetonyl-4-nitrobenzyl)-4-hydroxycoumarin, 3-(α-Acetonyl-4-nitrobenzyl)-4-hydroxycoumarin, 3-(α-Acetonyl-p-nitrobenzyl)-4-hydroxycoumarin, 3-(α-p-Nitrophenyl-β-acetylethyl)-4-hydroxycoumarin, 3-[2-Acetyl-1-(p-nitrophenyl)ethyl]-4-hydroxycoumarin, 3-[α-(4'-Nitrophenyl)-β-acetylethyl]-4-hydroxycoumarin, 3-[α-(p-Nitrophenol)-β-acetylethyl]-4-hydroxycoumarin, 4-Hydroxy-2-oxo-3-[3-oxo-1-(4-nitrophenyl)butyl]-2H-chromene, Acebron, Acenocoumarin, Acenokumarol, Acitrom, Ascumar, DL-3-(α-Acetonyl-4-nitrobenzyl)-4-hydroxycoumarin, G 23,350, G 23350, Minisintrom, Neo-Sintrom, Nicoumalone, Nitrowarfarin, Sincoumar, Sinkumar, Sinthrom, Sinthrome, Sintrom, Sintrom Mitis, Sintroma, Syncoumar, Syncumar, Syntrom, Trombostop, Zotil
IUPAC Name 4-hydroxy-3-[1-(4-nitrophenyl)-3-oxobutyl]chromen-2-one
SMILES CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3ccc(cc3)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H15NO6/c1-11(21)10-15(12-6-8-13(9-7-12)20(24)25)17-18(22)14-4-2-3-5-16(14)26-19(17)23/h2-9,15,22H,10H2,1H3
Formula Weight 353.0899
Chemical Name or Material Acenocoumarol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.