missing translation for 'onlineSavingsMsg'
Learn More

ACES, 0.5M buffer soln., pH 7.5, Thermo Scientific™

Product Code. 15473679
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473679 250 mL 250mL
15463679 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473679 Supplier Thermo Scientific Alfa Aesar Supplier No. J61011.AK
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N

Specifications

Color Colorless
Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material ACES buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.