missing translation for 'onlineSavingsMsg'
Learn More

Melford ACES

Product Code. 30018021
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Unit Size:
100g
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018021 5 kg 5kg
30018019 100 g 100g
30018020 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018021 Supplier Melford Supplier No. A225005000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zwitterionic buffer used in biochemistry and molecular biology research.

Zwitterionic buffer used in biochemistry and molecular biology research.

  • pKa (25°C): 6.88
  • The working pH range in aqueous solution is 6.1 - 7. 5
  • Absorbance 280nm (5% water): <0.02

Specifications

Chemical Name or Material ACES
CAS 7365-82-4
Quantity 5 kg
Molecular Formula C4H10N2O4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.