missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ACES, 99%

Product Code. 11454593
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454593 100 g 100g
11444593 25 g 25g
11464593 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11454593 Lieferant Thermo Scientific Chemicals Lieferanten-Nr. A11553.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol

ACES is used as a Good's buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ACES is used as a Good′s buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

Solubility
Soluble in water(18g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym aces, n-carbamoylmethyl taurine, n-2-acetamido-2-aminoethanesulfonic acid, 2-2-amino-2-oxoethyl amino ethanesulfonic acid, ethanesulfonic acid, 2-2-amino-2-oxoethyl amino, unii-i5spl5yvby, i5spl5yvby, 2-carbamoylmethylamino ethanesulfonic acid
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N

Spezifikation

Melting Point ∼275°C (decomposition)
Quantity 100 g
Beilstein 2253770
Merck Index 14,36
Solubility Information Soluble in water(18g/L).
Formula Weight 182.2
Percent Purity 99%
Chemical Name or Material ACES

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.