missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ACES, 99+%, for biochemistry

Product Code. 10741745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10741745 250 g Plastic bottle 250g
10183531 100 g Plastic bottle 100g
10117813 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10741745 Supplier Thermo Scientific Chemicals Supplier No. 327672500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACES, >-99%, C4H10N2O4S, CAS Number-7365-82-4, N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine, 1kg, 0.02 max. at 280nm, 0.04 max. at 260nm, (5% aq. soln.), (5% aq. soln.) at 0nm, 99+%, 100.0, 99.0, CHEBI:39062, 100ppm max., White, 230-908-4, 182.19, Biochemical | CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N

Specifications

Absorbance (5% aq. soln.), (5% aq. soln.) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
Absorbance Conditions (5% aq. soln.)
Melting Point 270.0°C to 295.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
pH 3.6 to 4.4 (1% soln. at 25°C)
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula H2NCOCH2NHCH2CH2SO3H
Packaging Plastic bottle
Merck Index 15, 37
Solubility Information Solubility in water: soluble (approx. 40g/L at 20 °C).
Formula Weight 182.19
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material ACES
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.