missing translation for 'onlineSavingsMsg'
Learn More

ACES (Fine White Crystals), Fisher BioReagents

Product Code. 10429123
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
10429123 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 10429123

Brand: Fisher BioReagents BP303100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
ACES biological buffer has a usable pH range of 6.1 to 7.5.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7365-82-4
Melting Point 270°C
Color White
Decomposition Information 293°C
pH 3.6 to 4.4
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥96 %
Identification Pass Test
Molecular Formula C4H10N2O4S
Linear Formula H2NCOCH2NHCH2CH2SO3H
Packaging Glass Bottle
Merck Index 15, 37
Synonym N-(2-Acetamido)-2-aminomethane Sulfonic Acid
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILES C(CS(=O)(=O)O)NCC(=O)N
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
Molecular Weight (g/mol) 182.194
PubChem CID 81832
ChEBI CHEBI:39062
Formula Weight 182.19
Percent Purity ≥96%
Chemical Name or Material ACES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.