missing translation for 'onlineSavingsMsg'
Learn More

ACES, JustPURE™, white powder, Fisher BioReagents

Product Code. 10819020
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10819020

Brand: Fisher BioReagents BP2935100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Absorbance 0.05 max. at 260nm, 0.05 max. at 280nm
CAS 7365-82-4
Melting Point 270°C
Color White
Decomposition Information 293°C
pH 3.6 to 4.4
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥99.0 %
Molecular Formula C4H10N2O4S
Packaging Poly Bottle
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid
Solubility Information Soluble in water
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILES C(CS(=O)(=O)O)NCC(=O)N
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
Molecular Weight (g/mol) 182.194
PubChem CID 81832
ChEBI CHEBI:39062
Formula Weight 182.2
Percent Purity ≥99%
Grade JustPURE™
Chemical Name or Material ACES
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.