missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen-d4 (major) (1.0 mg/mL in Methanol), TRC

Product Code. 30389360
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389360 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389360 Supplier Toronto Research Chemicals Supplier No. TRCKIT24715X1ML
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64315-36-2
Molecular Formula C8 2H4 H5 N O2
Quantity 5 x 1 mL
Synonym Paracetamol D4 (phenyl D4), Paracetamol D4, Paracetamol-d4, Acetaminophen D4, Acetaminophen-d4, Acetamide, N-(4-hydroxyphenyl-2,3,5,6-d4)-, 4-Hydroxy-2,3,5,6-tetradeuteroacetanilide, N-(4-Hydroxyphenyl-2,3,5,6-d4)acetamide
InChI Formula InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(NC(=O)C)c([2H])c([2H])c1O
IUPAC Name N-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetamide
Molecular Weight (g/mol) 155.19
Formula Weight 155.0884
Concentration or Composition (by Analyte or Components) Paracetamol-D4 1 mg/mL
Chemical Name or Material Paracetamol-D4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.