missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen, MedChemExpress

Product Code. 16440618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440618 5 g 5g
16408168 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440618 Supplier Medchem Express Supplier No. HY66005
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM; is a widely used antipyretic and analgesic agent. Acetaminophen is a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.

Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM

Specifications

Chemical Name or Material Acetaminophen
CAS 103-90-2
Color White
Purity Grade Notes Research
Molecular Formula C8H9NO2
Quantity 5 g
Synonym Paracetamol 4-Acetamidophenol 4'-Hydroxyacetanilide
Solubility Information DMSO : 250 mg/mL (1653.88 mM; Need ultrasonic) ∣H2O : 10 mg/mL (66.16 mM; Need ultrasonic)
SMILES O=C(C)NC1=CC=C(O)C=C1
Molecular Weight (g/mol) 151.16
Formula Weight 151.16
Percent Purity 98.79%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.