missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen, TRC

Product Code. 17910825
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910825 5 g 5g
17922305 1 g 1g
17923135 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17910825 Supplier Toronto Research Chemicals Supplier No. TRCA1612205G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-90-2
Molecular Formula C8 H9 N O2
Molecular Weight (g/mol) 151.16
Synonym Acetamide, N-(4-hydroxyphenyl)-, Acetanilide, 4'-hydroxy-, 4'-Hydroxyacetanilide, 4-(Acetylamino)phenol, 4-(N-Acetylamino)phenol, 4-Acetamidophenol, 4-Acetaminophenol, 4-Hydroxyacetanilide, APAP, Abensanil, Acamol, Acenol, Acenol (pharmaceutical), Acetagesic, Acetalgin, Acetaminofen, Acetaminophen, Algotropyl, Alpiny, Alvedon, Amadil, Anaflon, Anelix, Anhiba, Apamid, Apamide, Banesin, Ben-u-ron, Bickie-mol, Biocetamol, Calpol, Captin, Cetadol, Citramon P, Claratal, Clixodyne, Crocin, Dafalgan, Daga, Daphalgan, Datril, Dial-a-gesic, Dirox, Disprol, Doliprane, Dolprone, Duorol, Dymadon, Efferalgan, Endophy, Enelfa, Eneril, Eu-Med, Exdol, Febrilex, Febrilix, Febro-Gesic, Febrolin, Fendon, Fepanil, Finimal, Gattaphen T, Gelocatil, Hedex, Homoolan, Jin Gang, Korum, Lestemp, Liquagesic, Lonarid, Lyteca, Lyteca Syrup, Momentum, Multin, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, N-Acetyl-4-hydroxyaniline, N-Acetyl-p-aminophenol, NAPA, NAPA (analgesic), NAPAP, NSC 109028, NSC 3991, Napafen, Naprinol, Nebs, Nobedon, Ortensan, Pacemo, Pacemol, Paldesic, Pamol, Panadol, Panadol Actifast, Panadol Extend, Panaleve, Panasorb, Panets, Panodil, Paracetamol, Paracetamol DC, Paracetamole, Paralen, Paramol, Paraspen, Parelan, Parmol, Pasolind N, Pedric, Perfalgan, Phenaphen, Phendon, Pinex, Pinex (pharmaceutical), Prodafalgan, Puerxitong, Pyrinazine, Resfenol, Resprin, Rhodapop NCR, Salzone, Sara, Tabalgin, Tachipirina, Tapar, Temlo, Tempanal, Tempra, Tralgon, Tylenol, Valadol, Valgesic, Vick Pyrena, p-(Acetylamino)phenol, p-Aceaminophenol, p-Acetamidophenol, p-Acetoaminophen, p-Hydroxyacetanilide, panadeine
IUPAC Name N-(4-hydroxyphenyl)acetamide
SMILES CC(=O)Nc1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)
Formula Weight 151.0633
Percent Purity >95%
Chemical Name or Material Paracetamol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.