missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid-2-methylbutyl ester, Dr. Ehrenstorfer

Product Code. 30316588
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30316588

Brand: Dr. Ehrenstorfer DREC10016260

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic Acid-2-Methylbutyl Ester
CAS 624-41-9
Analyte or Component Names Acetic Acid-2-Methylbutyl Ester
Shipping Condition +20°C
Molecular Formula C7 H14 O2
Certifications/Compliance ISO/IEC 17025
Synonym Acetic Acid-2-Methylbutyl Ester, 1-Butanol, 2-methyl-, acetate (6CI, 7CI, 8CI, 9CI), 2-Methyl-1-butyl acetate, 2-Methylbutanol acetate, 2-Methylbutyl acetate
InChI Formula InChI=1S/C7H14O2/c1-4-6(2)5-9-7(3)8/h6H,4-5H2,1-3H3
SMILES CCC(C)COC(=O)C
IUPAC Name 2-methylbutyl acetate
Molecular Weight (g/mol) 130.185
Quantity 1 mL
Formula Weight 130.099 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.