missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid solution, Dilute (12%) EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12907153 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12907153 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12907153 Supplier Fisher Chemical Supplier No. RP 002 1000
  • ONLINE EXCLUSIVE PRICE
    90.30 €
    73.75 € / 1000mL
    Save 16.55 €
    18% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 121-17-5
Analyte or Component Names Acetic Acid Solution (Dilute, 12%)
Molecular Formula C7H3ClF3NO2
Concentration 1.0 N
MDL Number MFCD00007084
InChI Key TZGFQIXRVUHDLE-UHFFFAOYSA-N
Type Mass Spectrometry Standards
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
Element Acetic Acid
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 225.55
PubChem CID 8462
Quantity 1000 mL
Grade Eur Ph Ref.: 1000402
Chemical Name or Material Aminohippuric acid reagent.
Show More Show Less
compliance-icons
Product Identifier
  • Acetic acid,dilute (12%) European Pharmacopoeia Ref: 1000402. (RP 002)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.