missing translation for 'onlineSavingsMsg'
Learn More

Acetochlor SAA 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30326562
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326562 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326562 Supplier Dr. Ehrenstorfer Supplier No. DREA10018450AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetochlor-sulfinyl-acetic acid
CAS 618113-86-3
Analyte or Component Names Acetochlor-sulfinyl-acetic acid
Shipping Condition +20°C
Molecular Formula C16 H23 N O5 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Acetic acid, [[2-[(ethoxymethyl)(2-ethyl-6-methylphenyl)amino]-2-oxoethyl]sulfinyl]- (9CI), 2-[[2-[(Ethoxymethyl)(2-ethyl-6-methylphenyl)amino]-2-oxoethyl]sulfinyl]acetic acid, Acetochlor-sulfinyl-acetic acid, Acetochlor SAA
InChI Formula InChI=1S/C16H23NO5S/c1-4-13-8-6-7-12(3)16(13)17(11-22-5-2)14(18)9-23(21)10-15(19)20/h6-8H,4-5,9-11H2,1-3H3,(H,19,20)
SMILES CCOCN(C(=O)CS(=O)CC(=O)O)c1c(C)cccc1CC
IUPAC Name 2-[2-[N-(ethoxymethyl)-2-ethyl-6-methyl-anilino]-2-oxo-ethyl]sulfinylacetic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 341.42
Quantity 1 mL
Formula Weight 341.1297 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.