missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-CoA Carboxylase Antibody (SR1412), Novus Biologicals™

Rabbit Monoclonal Antibody

349.00 € - 449.00 €

Specifications

Antigen Acetyl-CoA Carboxylase
Clone SR1412
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18664315
View Documents
Novus Biologicals
NBP3-22652-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18692565
View Documents
Novus Biologicals
NBP3-22652-50ul
50 μg
349.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acetyl-CoA Carboxylase Monoclonal antibody specifically detects Acetyl-CoA Carboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Acetyl-CoA Carboxylase
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
IgG
Affinity purified
SR1412
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
A synthesized peptide derived from human Acetyl-CoA Carboxylase (Uniprot #: Q13085, O00763)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.