missing translation for 'onlineSavingsMsg'
Learn More

Acetylcedrene (~80%), TRC

Product Code. 30012624
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012624 25 g 25g
30010042 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012624 Supplier Toronto Research Chemicals Supplier No. TRCA17198525G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32388-55-9
Molecular Formula C17 H26 O
Molecular Weight (g/mol) 246.39
Synonym alpha-Acetylcedrene, Ethanone, 1-(2,3,4,7,8,8a-hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl)-, [3R-(3α,3aβ,7β,8aα)]-, 1-[(3R,3aR,7R,8aS)-2,3,4,7,8,8a-Hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl]ethanone, 1H-3a,7-Methanoazulene, ethanone deriv., 1-((3R,3AR,7R,8aS)-3,6,8,8-tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl)ethan-1-one, 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, Acetyl-α-cedrene, Acetylcedrene, Lixetone, Vertofix, Ethanone, 1-[(3R,3aR,7R,8aS)-2,3,4,7,8,8a-hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl]-, Acetyl-alpha-cedrene, 1-[(3R,3aR,7R,8aS)-2,3,4,7,8,8a-Hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl]ethanone, 1H-3a,7-Methanoazulene, ethanone deriv., 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, Acetyl-α-cedrene, Acetylcedrene, Lixetone, Vertofix
IUPAC Name 1-[(1R,2R,5S,7R)-2,6,6,8-tetramethyl-9-tricyclo[5.3.1.01,5]undec-8-enyl]ethanone
SMILES C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC(=C3C)C(=O)C

Specifications

Analyte or Component Names Acetyl-alpha-cedrene
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H26O/c1-10-6-7-15-16(4,5)14-9-17(10,15)8-13(11(14)2)12(3)18/h10,14-15H,6-9H2,1-5H3/t10-,14+,15+,17+/m1/s1
Solution Type Neat
Industry Type Forensics
Quantity 25 g
Formula Weight 246.1984
Chemical Name or Material Acetyl-alpha-cedrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.