missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine chloride, MedChemExpress

Product Code. 16452119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452119 1 g 1g
16440858 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452119 Supplier Medchem Express Supplier No. HYB0282
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Acetylcholine chloride (ACh chloride), a neurotransmitter, is a potent cholinergic agonist. Acetylcholine chloride is a modulator of the activity of dopaminergic (DAergic) neurons through the stimulation of nicotinic acetylcholine receptors (nAChRs). Acetylcholine chloride inhibits p53 mutant peptide aggregation in vitro.

Acetylcholine chloride (ACh chloride), a neurotransmitter, is a potent cholinergic agonist. Acetylcholine chloride is a modulator of the activity of dopaminergic (DAergic) neurons through the stimulation of nicotinic acetylcholine receptors (nAChRs). Acetylcholine chloride inhibits p53 mutant peptide aggregation in vitro.

Specifications

Chemical Name or Material Acetylcholine chloride
Physical Form Solid
CAS 60-31-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C7H16ClNO2
For Use With (Application) Cancer-programmed cell death
Synonym ACh chloride
Solubility Information H2O : 100 mg/mL (550.48 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (344.05 mM; Need ultrasonic)
SMILES O=C(OCC[N+](C)(C)C)C.[Cl-]
Molecular Weight (g/mol) 181.66
Formula Weight 181.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.