missing translation for 'onlineSavingsMsg'
Learn More

Acetylcysteine, MedChemExpress

Product Code. 16488418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488418 10 g 10g
16460828 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488418 Supplier Medchem Express Supplier No. HYB0215
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acetylcysteine (N-Acetylcysteine) is a mucolytic agent which reduces the thickness of the mucus. Acetylcysteine is a ROS inhibitor. Acetylcysteine is a cysteine precursor, prevents hemin-induced ferroptosis by neutralizing toxic lipids generated by arachidonate-dependent activity of 5-lipoxygenases. Acetylcysteine induces cell apoptosis. Acetylcysteine also has anti-influenza virus activities.

Acetylcysteine (N-Acetylcysteine) is a mucolytic agent which reduces the thickness of the mucus. Acetylcysteine is a ROS inhibitor. Acetylcysteine is a cysteine precursor, prevents hemin-induced ferroptosis by neutralizing toxic lipids generated by arachidonate-dependent activity of 5-lipoxygenases. Acetylcysteine induces cell apoptosis. Acetylcysteine also has anti-influenza virus activities.

Specifications

Chemical Name or Material Acetylcysteine
Physical Form Solid
CAS 616-91-1
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C5H9NO3S
For Use With (Application) Neuroscience-Neurodegeneration
Synonym N-Acetylcysteine N-Acetyl-L-cysteine NAC
Solubility Information DMSO : ≥ 130 mg/mL (796.62 mM) ∣H2O : 50 mg/mL (306.39 mM; Need ultrasonic)
SMILES SC[C@@H](C(O)=O)NC(C)=O
Molecular Weight (g/mol) 163.19
Formula Weight 163.19
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.