missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic acid, 99%, Thermo Scientific Chemicals

Artikelnummer. 10318893 Alle ansehen Thermo Scientific Produkte
Click to view available options
Quantity:
50 g
500 g
2.5 kg
Packaging:
Plastic bottle
Packungsgröße:
2.5kg
500g
50g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 10318893

Marke: Thermo Scientific 158180025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Acetylsalicylic acid, aspirin, CAS# 50-78-2, is a non-steroidal anti-inflammatory that is used widely as an analgesic and antipyretic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Acetylsalicylic acid is a common non-steroidal anti-inflammatory (NSAID) and
• Acetylsalicylic acid works by inhibiting cyclooxygenases, reducing prostaglandin production and the release of pyrogens


Applications
• Acetylsalicylic acid is useful for research on fever, postoperative pain, blood clots, and inflammation
• Acetylsalicylic acid may also be used in the study of cancer prevention
• Intended for research use only

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Spezifikation

CAS Min % 98.5
CAS Max % 100.0
Melting Point 136.0°C to 140.0°C
Color White
Density 1.3500g/mL
Flash Point 250°C
Loss on Drying 0.5% max. (1 g) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula 2-(CH3CO2)C6H4CO2H
Quantity 2.5 kg
Beilstein 10, 67
Merck Index 15, 841
Specific Gravity 1.35
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: 1g in 5mL alcohol,17mL chloroform,,and 10-15 mL ether,slightly soluble in anhydrous ether,decomposes in boiling water,alkali
Formula Weight 180.16
Percent Purity 99%
Physical Form Granular Powder
Chemical Name or Material Acetylsalicylic acid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Acetylsalicylic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
Gefahrenhinweis
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt