missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acid Fuchsin sodium salt

Product Code. 11475856
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475856 500 g 500g
11455856 25 g 25g
11465856 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475856 Supplier Thermo Scientific Chemicals Supplier No. B22222.36
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3244-88-0 | C20H19N3Na2O9S3 | 587.544 g/mol

Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

Solubility
Soluble in water (1 mg/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3244-88-0
Molecular Formula C20H19N3Na2O9S3
Molecular Weight (g/mol) 587.544
MDL Number MFCD00013286
InChI Key VCEFFPMSOGDMJX-UHFFFAOYSA-N
Synonym acid fuchsin, acidal fuchsine, acid fuchsine, acidal magenta, fuchsine acid, fuchsin acid, p-fuchsine acid, acid fuchsine n, acid fuchsine o, acid fuchsine s
PubChem CID 131852436
IUPAC Name 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
SMILES CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]

Specifications

Quantity 500 g
Beilstein 4111656
Merck Index 14,107
Solubility Information Soluble in water (1mg/ml).
Formula Weight 585.53
Chemical Name or Material Acid Fuchsin sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.