missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Surrogate Standard, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 2,4,6-Tribromophenol | 118-79-6 | 2000μg/mL |
| 2-Chlorophenol-3,4,5,6-d4 | 93951-73-6 | 2000μg/mL |
| 2-Fluorophenol | 367-12-4 | 2000μg/mL |
| Phenol-d6 | 13127-88-3 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Acid Surrogate Standard |
| Packaging | 5mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS | 118-79-6, 93951-73-6, 367-12-4, 13127-88-3 |
| Analyte or Component Names | 2,4,6-Tribromophenol |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Element | 2,4,6-Tribromophenol |
| Solvent or Matrix | Methanol |
| Quantity | 5 mL |
| Concentration or Composition (by Analyte or Components) | 2000μg/mL |
Safety and Handling
Identifiant produit
- Acid Surrogate Standard, 5 mL
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol
Product intended for laboratory use only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu