missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acifluorfen-methyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Acifluorfen methyl ester |
| CAS | 50594-67-7 |
| Analyte or Component Names | Acifluorfen methyl ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H9 Cl F3 N O5 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C15H9ClF3NO5/c1-24-14(21)10-7-9(3-4-12(10)20(22)23)25-13-5-2-8(6-11(13)16)15(17,18)19/h2-7H,1H3 |
| SMILES | COC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-] |
| IUPAC Name | methyl 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?