missing translation for 'onlineSavingsMsg'
Learn More

Aclidinium Bromide, MedChemExpress

Product Code. 16476396
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476396 100 mg 100mg
16475968 10 mg 10mg
16442117 50 mg 50mg
16418597 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476396 Supplier Medchem Express Supplier No. HY14144
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aclidinium Bromide (LAS 34273; LAS-W 330) is a long-acting, inhaled muscarinic antagonist. Aclidinium Bromide has the potential for chronic obstructive pulmonary disease (COPD) research.

Aclidinium Bromide (LAS 34273

Specifications

Chemical Name or Material Aclidinium Bromide
CAS 320345-99-1
Color Pink
Purity Grade Notes Research
Molecular Formula C26H30BrNO4S2
Quantity 100 mg
Synonym LAS 34273 LAS-W 330
Solubility Information DMSO : 33.33 mg/mL (59.04 mM; Need ultrasonic)
SMILES O=C(O[C@H]1C[N+]2(CCCOC3=CC=CC=C3)CCC1CC2)C(C4=CC=CS4)(O)C5=CC=CS5.[Br-]
Molecular Weight (g/mol) 564.55
Formula Weight 564.55
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.