missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACN9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06162-50ul
This item is not returnable.
View return policy
Description
ACN9 Polyclonal antibody specifically detects ACN9 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| ACN9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| ACN9 homolog (S. cerevisiae), DC11, protein ACN9 homolog, mitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human ACN9 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57001 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction