missing translation for 'onlineSavingsMsg'
Learn More

ACOT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16832135 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16832135 150 μL 150µL
16842135 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16832135 Supplier Proteintech Supplier No. 229361AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ACOT6 gene ontology annotations related to this gene include acyl-CoA metabolic process; fatty acid metabolic process.

Specifications

Antigen ACOT6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACOT6
Gene Accession No. Q32Q92, Q3I5F7
Gene Alias ACOT6, acyl CoA thioesterase 6, c14_5530, C14orf42
Gene Symbols ACOT6
Host Species Rabbit
Immunogen ACOT6 Fusion Protein Ag18981
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217700, 299193, 641372
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.