missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97895-100ul
This item is not returnable.
View return policy
Description
ACOX1 Polyclonal antibody specifically detects ACOX1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ACOX1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| ACOXEC 1.3.3.6, acyl-CoA oxidase 1, palmitoyl, acyl-CoA oxidase, straight-chain, acyl-Coenzyme A oxidase 1, palmitoyl, AOX, PALMCOX, Palmitoyl-CoA oxidase, peroxisomal acyl-coenzyme A oxidase 1, peroxisomal fatty acyl-CoA oxidase, SCOXMGC1198, Straight-chain acyl-CoA oxidase | |
| Produced in rabbits immunized with E. coli-derived Human ACOX1 fragment. | |
| 100 μg | |
| Lipid and Metabolism | |
| 51 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction