missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Acridine Orange hydrochloride
Specifications
Specifications
| Content And Storage | 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen) |
| Product Type | Dye Reagents |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 50 mg/mL (165.67 mM; ultrasonic and heat to 60°C) ∣ DMSO : 25 mg/mL (82.83 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 301.81 g/mol |
| CAS | 65-61-2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?