missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein Antibody (10A10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59359APC
This item is not returnable.
View return policy
Description
Acrolein Monoclonal specifically detects Acrolein in Non-species specific samples. It is validated for Flow Cytometry.
Specifications
| Acrolein | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Non-species specific | |
| Purified |
| Flow Cytometry | |
| 10A10 | |
| Flow Cytometry | |
| Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde | |
| Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μL | |
| Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction