missing translation for 'onlineSavingsMsg'
Learn More

ACTA2/smooth muscle actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16843014 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16843014 150 μL 150µL
16853014 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16843014 Supplier Proteintech Supplier No. 143951AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody may cross-react with other isoforms of actins.

Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predomitly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.

Specifica

Antigen ACTA2/smooth muscle actin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACTA2
Gene Accession No. P62736, P62737, P62738
Gene Alias a SMA, AAT6, ACTA2, ACTA2 Smooth muscle Actin, Actin, aortic smooth muscle, Actin, aortic smooth muscle Actin, ACTSA, ACTVS, Alpha actin 2, Alpha SMA, a-SMA, alpha SMA
Gene Symbols ACTA2
Host Species Rabbit
Immunogen ACTA2/smooth muscle actin Fusion Protein Ag5886
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11475, 59, 81633
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.