missing translation for 'onlineSavingsMsg'
Learn More
Learn More
actin-related protein 2/3 complex subunit 1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | actin-related protein 2/3 complex subunit 1B |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646845
|
Novus Biologicals
NBP2-99248-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663576
|
Novus Biologicals
NBP2-99248-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
actin-related protein 2/3 complex subunit 1B Polyclonal antibody specifically detects actin-related protein 2/3 complex subunit 1B in Human samples. It is validated for Western BlotSpecifications
| actin-related protein 2/3 complex subunit 1B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| actin related protein 2/3 complex, subunit 1B (41 kD), actin related protein 2/3 complex, subunit 1B, 41kDa, actin-related protein 2/3 complex subunit 1B, ARC41p41-ARCp40-ARC, Arp2/3 complex 41 kDa subunit, ARP2/3 protein complex subunit p41 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human actin-related protein 2/3 complex subunit 1B. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 10095 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title